Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp6v0e1
Entrez Gene ID
100769759
Gene Name
ATPase, H+ transporting, lysosomal 9kDa, V0 subunit e1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613815: 1143707 - 1169619 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500820.2 XP_003500868.1 3 - 4 406 71
GenBank mRNA JI889023.1 - 3 - 4 379 71

View Full Product Details