Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igj
Entrez Gene ID
100774033
Gene Name
immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614817: 70939 - 78275 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510044.2 XP_003510092.1 1 - 2 124 79
GenBank mRNA JI896010.1 - 1 - 2 194 79

View Full Product Details