Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt13
Entrez Gene ID
100758976
Gene Name
keratin 13
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615080: 173432 - 177333 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511328.1 XP_003511376.1 7 - 8 1255 72

View Full Product Details