Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spaca5
Entrez Gene ID
100766432
Gene Name
sperm acrosome associated 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614457: 226058 - 229015 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507950.2 XP_003507998.1 3 - 4 382 100

View Full Product Details