Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgpp1
Entrez Gene ID
100766871
Gene Name
sphingosine-1-phosphate phosphatase 1
Gene Aliases
I79_005352
Chromosome Location
Chr.Un|NW_003613728: 1521811 - 1548615 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499319.2 XP_003499367.1 1 - 2 46 78

View Full Product Details