Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100771726
Entrez Gene ID
100771726
Gene Name
chromosome unknown open reading frame, human C9orf96
Gene Aliases
I79_020325
Chromosome Location
Chr.Un|NW_003615220: 95724 - 118077 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652260.1 XP_007650450.1 16 - 17 1898 95

View Full Product Details