Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmaip1
Entrez Gene ID
100755393
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Gene Aliases
I79_014604
Chromosome Location
Chr.Un|NW_003614319: 496049 - 501321 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647555.1 XP_007645745.1 3 - 4 400 63

View Full Product Details