Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb17
Entrez Gene ID
100759666
Gene Name
ankyrin repeat and SOCS box containing 17
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613622: 3488927 - 3499004 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496442.2 XP_003496490.1 2 - 3 676 104

View Full Product Details