Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl20
Entrez Gene ID
100759818
Gene Name
methyltransferase like 20
Gene Aliases
I79_012677
Chromosome Location
Chr.Un|NW_003614152: 140216 - 155750 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505481.2 XP_003505529.1 2 - 3 504 106
XM_007645985.1 XP_007644175.1 2 - 3 504 106
GenBank mRNA JI873034.1 - 2 - 3 516 106

View Full Product Details