Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cxcl6
Entrez Gene ID
100759933
Gene Name
chemokine (C-X-C motif) ligand 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614806: 256554 - 257617 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510008.2 XP_003510056.1 3 - 4 331 85

View Full Product Details