Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Magix
Entrez Gene ID
100759147
Gene Name
MAGI family member, X-linked
Gene Aliases
I79_012256
Chromosome Location
Chr.Un|NW_003614122: 361487 - 367617 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505084.2 XP_003505132.1 1 - 2 48 57

View Full Product Details