Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl54
Entrez Gene ID
100763792
Gene Name
mitochondrial ribosomal protein L54
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614244: 597210 - 599745 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506450.2 XP_003506498.1 2 - 3 329 83
GenBank mRNA JI881469.1 - 1 - 2 165 83
JP059163.1 - 1 - 2 165 83

View Full Product Details