Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ebln2
Entrez Gene ID
103159492
Gene Name
endogenous Bornavirus-like nucleoprotein 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613950: 792321 - 792846 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643820.1 XP_007642010.1 2 - 3 251 108

View Full Product Details