Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Timd4
Entrez Gene ID
100763899
Gene Name
T-cell immunoglobulin and mucin domain containing 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614759: 201650 - 229086 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650353.1 XP_007648543.1 2 - 3 396 83

View Full Product Details