Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prx
Entrez Gene ID
100763689
Gene Name
periaxin
Gene Aliases
I79_012400
Chromosome Location
Chr.Un|NW_003614129: 535535 - 549286 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505240.2 XP_003505288.1 3 - 4 407 68

View Full Product Details