Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc31a2
Entrez Gene ID
100754404
Gene Name
solute carrier family 31 (copper transporter), member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613973: 1355407 - 1367419 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503464.2 XP_003503512.1 2 - 3 170 78
GenBank mRNA JI886384.1 - 2 - 3 79
JP042821.1 - 2 - 3 135 78

View Full Product Details