Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fgf5
Entrez Gene ID
100763237
Gene Name
fibroblast growth factor 5
Gene Aliases
I79_017276
Chromosome Location
Chr.Un|NW_003614663: 386252 - 405884 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509163.1 XP_003509211.1 1 - 2 340 76

View Full Product Details