Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a18
Entrez Gene ID
100753692
Gene Name
membrane-spanning 4-domains, subfamily A, member 18
Gene Aliases
I79_023662
Chromosome Location
Chr.Un|NW_003616604: 2148 - 13069 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654572.1 XP_007652762.1 1 - 2 455 119

View Full Product Details