Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Odf4
Entrez Gene ID
100752383
Gene Name
outer dense fiber of sperm tails 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613643: 1016735 - 1021230 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497222.1 XP_003497270.1 4 - 5 649 78

View Full Product Details