Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sult1c2
Entrez Gene ID
100759668
Gene Name
sulfotransferase family, cytosolic, 1C, member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616461: 31197 - 42459 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514252.1 XP_003514300.1 4 - 5 516 106

View Full Product Details