Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arsh
Entrez Gene ID
100773352
Gene Name
arylsulfatase family, member H
Gene Aliases
I79_018039
Chromosome Location
Chr.Un|NW_003614774: 78231 - 87623 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650487.1 XP_007648677.1 1 - 2 41 71

View Full Product Details