Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rassf7
Entrez Gene ID
100774797
Gene Name
Ras association (RalGDS/AF-6) domain family (N-terminal) member 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614763: 71929 - 73951 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509754.2 XP_003509802.1 3 - 4 1034 103

View Full Product Details