Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc74b
Entrez Gene ID
100757786
Gene Name
coiled-coil domain containing 74B
Gene Aliases
I79_009528
Chromosome Location
Chr.Un|NW_003613919: 1557944 - 1562551 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502842.2 XP_003502890.1 7 - 8 715 57

View Full Product Details