Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmrt2
Entrez Gene ID
100756035
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003618188: 11899 - 17664 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515478.2 XP_003515526.1 2 - 3 650 107

View Full Product Details