Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrtm1
Entrez Gene ID
103159741
Gene Name
leucine-rich repeats and transmembrane domains 1
Gene Aliases
I79_001461
Chromosome Location
Chr.Un|NW_003613611: 2596305 - 2603562 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647207.1 XP_007645397.1 3 - 4 711 71

View Full Product Details