Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxc10
Entrez Gene ID
100764281
Gene Name
homeobox C10
Gene Aliases
I79_014805
Chromosome Location
Chr.Un|NW_003614339: 80831 - 85695 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507190.2 XP_003507238.2 1 - 2 838 65

View Full Product Details