Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt71
Entrez Gene ID
100754630
Gene Name
keratin 71
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615420: 117468 - 126414 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512325.1 XP_003512373.1 8 - 9 1357 92

View Full Product Details