Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbx10
Entrez Gene ID
100763900
Gene Name
T-box 10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614797: 102641 - 105558 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650602.1 XP_007648792.1 6 - 7 862 71

View Full Product Details