Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhod
Entrez Gene ID
100760521
Gene Name
ras homolog family member D
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614834: 438812 - 450119 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650789.1 XP_007648979.1 3 - 4 369 75

View Full Product Details