Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt72
Entrez Gene ID
100752072
Gene Name
keratin 72
Gene Aliases
I79_021072, Krt72-ps
Chromosome Location
Chr.Un|NW_003615420: 156442 - 166683 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512317.1 XP_003512365.1 8 - 9 1120 70

View Full Product Details