Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Isx
Entrez Gene ID
100756409
Gene Name
intestine-specific homeobox
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617336: 5636 - 26933 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515099.1 XP_003515147.1 1 - 2 218 61

View Full Product Details