Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bok
Entrez Gene ID
100773011
Gene Name
BCL2-related ovarian killer
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613726: 1249372 - 1259574 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499263.2 XP_003499311.1 3 - 4 514 109

View Full Product Details