Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcrlb
Entrez Gene ID
100771895
Gene Name
Fc receptor-like B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614301: 551519 - 557160 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506877.1 XP_003506925.1 5 - 6 855 77

View Full Product Details