Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35e4
Entrez Gene ID
100757148
Gene Name
solute carrier family 35, member E4
Gene Aliases
I79_017100
Chromosome Location
Chr.Un|NW_003614628: 374491 - 382862 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509017.2 XP_003509065.1 1 - 2 1240 63
XM_007649654.1 XP_007647844.1 2 - 3 1581 63
GenBank mRNA JI876184.1 - 1 - 2 173 63

View Full Product Details