Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdr42e1
Entrez Gene ID
100752182
Gene Name
short chain dehydrogenase/reductase family 42E, member 1
Gene Aliases
I79_000292
Chromosome Location
Chr.Un|NW_003613583: 5893187 - 5895190 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495129.1 XP_003495177.1 1 - 2 75 96

View Full Product Details