Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam69c
Entrez Gene ID
100775103
Gene Name
family with sequence similarity 69, member C
Gene Aliases
I79_021137
Chromosome Location
Chr.Un|NW_003615437: 2891 - 16133 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512381.2 XP_003512429.1 2 - 3 157 66

View Full Product Details