Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Htr3b
Entrez Gene ID
100764440
Gene Name
5-hydroxytryptamine (serotonin) receptor 3B, ionotropic
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617137: 2295 - 25849 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514938.2 XP_003514986.2 3 - 4 307 109

View Full Product Details