Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gbx1
Entrez Gene ID
100769092
Gene Name
gastrulation brain homeobox 1
Gene Aliases
I79_007685
Chromosome Location
Chr.Un|NW_003613842: 969775 - 998866 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641982.1 XP_007640172.1 2 - 3 146 82

View Full Product Details