Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dbndd1
Entrez Gene ID
100759269
Gene Name
dysbindin (dystrobrevin binding protein 1) domain containing 1
Gene Aliases
I79_000189
Chromosome Location
Chr.Un|NW_003613583: 26939 - 34064 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650635.1 XP_007648825.1 2 - 2 923 88

View Full Product Details