Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem51
Entrez Gene ID
100766562
Gene Name
transmembrane protein 51
Gene Aliases
I79_024090
Chromosome Location
Chr.Un|NW_003616957: 62050 - 68780 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514775.2 XP_003514823.1 2 - 2 618 110

View Full Product Details