The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005689436.3 | XP_005689493.1 | 1 - 2 | 502 | 74 |
GenBank mRNA | JO442347.2 | - | 1 - 2 | 512 | 74 |