Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C10H15orf41
Entrez Gene ID
102171792
Gene Name
chromosome 10 C15orf41 homolog
Gene Aliases
-
Chromosome Location
Chr.10: 70560870 - 70793830 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005685467.3 XP_005685524.1 2 - 3 344 71
XM_013966954.2 XP_013822408.1 2 - 3 344 71
XM_013966957.2 XP_013822411.1 3 - 4 383 71
XM_018054491.1 XP_017909980.1 2 - 3 344 71

View Full Product Details