Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIRAP
Entrez Gene ID
102184789
Gene Name
TIR domain containing adaptor protein
Gene Aliases
-
Chromosome Location
Chr.29: 28968489 - 28986263 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043302.1 XP_017898791.1 5 - 6 1288 74
XM_018043303.1 XP_017898792.1 5 - 6 1283 74
XM_018043304.1 XP_017898793.1 5 - 6 1089 74
XM_018043305.1 XP_017898794.1 4 - 5 991 74

View Full Product Details