Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCKBR
Entrez Gene ID
102186886
Gene Name
cholecystokinin B receptor
Gene Aliases
-
Chromosome Location
Chr.15: 35572263 - 35583453 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059455.1 XP_017914944.1 3 - 4 753 101
XM_018059456.1 XP_017914945.1 3 - 4 942 101
XM_018059457.1 XP_017914946.1 4 - 5 648 101
GenBank mRNA KC767906.1 - 2 - 3 403 101

View Full Product Details