Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUCKS1
Entrez Gene ID
102183256
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Gene Aliases
-
Chromosome Location
Chr.16: 2862031 - 2894289 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060062.1 XP_017915551.1 2 - 3 331 88
XM_018060064.1 XP_017915553.1 2 - 3 332 88

View Full Product Details