Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHFPL4
Entrez Gene ID
102185875
Gene Name
LHFPL tetraspan subfamily member 4
Gene Aliases
-
Chromosome Location
Chr.22: 16905085 - 16938584 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973450.2 XP_013828904.1 2 - 3 1757 68

View Full Product Details