Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASL12
Entrez Gene ID
102182040
Gene Name
RAS like family 12
Gene Aliases
-
Chromosome Location
Chr.10: 89261526 - 89277905 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053658.1 XP_017909147.1 4 - 5 569 86
GenBank mRNA JO566132.2 - - 690 86
KA308244.1 - 2 - 3 1761 86

View Full Product Details