Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTHRC1
Entrez Gene ID
102188151
Gene Name
collagen triple helix repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 21035019 - 21051028 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013969243.2 XP_013824697.1 3 - 4 765 82

View Full Product Details