Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP3
Entrez Gene ID
102168333
Gene Name
A-kinase anchoring protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 104412263 - 104440801 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048610.1 XP_017904099.1 3 - 4 407 71
XM_018048611.1 XP_017904100.1 4 - 5 496 71

View Full Product Details