Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EFNA3
Entrez Gene ID
102177853
Gene Name
ephrin A3
Gene Aliases
-
Chromosome Location
Chr.3: 104612091 - 104621526 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KA344465.1 - 3 - 4 1637 71

View Full Product Details